missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dihydroxytoluene monohydrate, 97%

Catalog No. p-7080645
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1044006 5 g
AAA1044014 25 g
AAA1044022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1044006 Supplier Thermo Scientific Chemicals Supplier No. A1044006
Only null left

CAS: 6153-39-5 | C7H10O3 | 142.154 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6153-39-5
Molecular Formula C7H10O3
Molecular Weight (g/mol) 142.154
MDL Number MFCD00149092
InChI Key NBKPNAMTHBIMLA-UHFFFAOYSA-N
Synonym 5-methylbenzene-1,3-diol hydrate, orcinol monohydrate, 5-methylresorcinol monohydrate, 3,5-dihydroxytoluene monohydrate, orcinol hydrate, resorcinol, 5-methyl-, monohydrate, 1,3-benzenediol, 5-methyl-, monohydrate, 1,3-dihydroxy-5-methylbenzene monohydrate, acmc-209mtl, 5-methylresorcinol hydrate
PubChem CID 3083941
IUPAC Name 5-methylbenzene-1,3-diol;hydrate
SMILES CC1=CC(=CC(=C1)O)O.O

Specifications

Melting Point 56°C to 60°C
Boiling Point 147°C (5 mmHg)
Quantity 5 g
Beilstein 5103474
Sensitivity Air Sensitive
Merck Index 14,6864
Formula Weight 142.16 (124.14 Anhydrous)
Percent Purity 97%
Chemical Name or Material 3,5-Dihydroxytoluene monohydrate
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 207-984-2
RTECSNumber VH2100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only