missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dihydroxybenzonitrile, 98%

Catalog No. AAL0564203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0564203 1 g
AAL0564206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0564203 Supplier Thermo Scientific Chemicals Supplier No. L0564203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 19179-36-3 | C7H5NO2 | 135.12 g/mol

3,5-Dihydroxybenzonitrile is used in the preparation of 3-hydroxy-5-methoxybenzonitrile using potassium carbonate as a reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dihydroxybenzonitrile is used in the preparation of 3-hydroxy-5-methoxybenzonitrile using potassium carbonate as a reagent.

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents.

Chemical Identifiers

CAS 19179-36-3
Molecular Formula C7H5NO2
Molecular Weight (g/mol) 135.12
MDL Number MFCD00016453
InChI Key ABHOEQJNEOMTEK-UHFFFAOYSA-N
Synonym benzonitrile, 3,5-dihydroxy, benzonitrile,3,5-dihydroxy, 3,5-dihydroxy benzonitrile, 5-cyanoresorcinol, a-resorcylonitrile, pubchem13116, acmc-1bouo
PubChem CID 87952
IUPAC Name 3,5-dihydroxybenzonitrile
SMILES OC1=CC(=CC(O)=C1)C#N

Specifications

Melting Point 185°C to 190°C
Quantity 1 g
UN Number UN3439
Beilstein 6322491
Solubility Information Soluble in water.
Formula Weight 135.12
Percent Purity 98%
Chemical Name or Material 3,5-Dihydroxybenzonitrile
Hazard Category H302+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 242-859-6
TSCA No

RUO – Research Use Only