missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dihydroxybenzhydrazide, 98%

Catalog No. p-7045648
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1147106 5 g
AAL1147114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1147106 Supplier Thermo Scientific Chemicals Supplier No. L1147106
Only null left

CAS: 7732-32-3 | C7H8N2O3 | 168.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7732-32-3
Molecular Formula C7H8N2O3
Molecular Weight (g/mol) 168.15
MDL Number MFCD00017066
InChI Key WGXWEXNJRZMIPT-UHFFFAOYSA-N
Synonym 3,5-dihydroxybenzhydrazide, 3,5-dihydroxy-benzhydrazide, 5-???benzene-1,3-diol, #, 3,5-dihydroxybenzoic hydrazide, 3,5-bis oxidanyl benzohydrazide, benzoic acid,3,5-dihydroxy-, hydrazide, 3,5-dihydroxy-benzoic acid hydrazide
PubChem CID 586309
IUPAC Name 3,5-dihydroxybenzohydrazide
SMILES NNC(=O)C1=CC=C(O)C(O)=C1

Specifications

Melting Point 256°C to 257°C
Quantity 5 g
Beilstein 2642968
Formula Weight 168.15
Percent Purity 98%
Chemical Name or Material 3,5-Dihydroxybenzhydrazide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only