missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dihydroxy-4-methylbenzoic Acid 98.0+%, TCI America™

Catalog No. D47121G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47121G 1 g
D47125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D47121G Supplier TCI America Supplier No. D47121G

Chemical Identifiers

CAS 28026-96-2
Molecular Formula C8H7O4
Molecular Weight (g/mol) 167.14
MDL Number MFCD00002514
InChI Key KMRRXSZDSGYLCD-UHFFFAOYSA-M
Synonym 3,5-dihydroxy-p-toluic acid, benzoic acid, 3,5-dihydroxy-4-methyl, 4-methyl-3,5-dihydroxy benzoic acid, 3,5-dihydroxy-4-methyl-benzoic acid, pubchem3868, acmc-20a50j, paragos 440034, 3,5-dihydroxy4-methylbenzoic acid, 3,5-dihydroxy-4-methyl benzoic acid, benzoic acid,3,5-dihydroxy-4-methyl
PubChem CID 2748045
IUPAC Name 3,5-dihydroxy-4-methylbenzoate
SMILES CC1=C(O)C=C(C=C1O)C([O-])=O

Specifications

Melting Point 269°C
Color White-Yellow
Quantity 1 g
Formula Weight 168.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dihydroxy-4-methylbenzoic Acid