missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dihydroxy-4-methylbenzoic acid, 97%

Catalog No. p-7024473
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1919603 1 g
AAA1919606 5 g
AAA1919614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1919603 Supplier Thermo Scientific Chemicals Supplier No. A1919603
Only null left

CAS: 28026-96-2 | C8H7O4 | 167.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 28026-96-2
Molecular Formula C8H7O4
Molecular Weight (g/mol) 167.14
MDL Number MFCD00002514
InChI Key KMRRXSZDSGYLCD-UHFFFAOYSA-M
PubChem CID 2748045
IUPAC Name 3,5-dihydroxy-4-methylbenzoic acid
SMILES CC1=C(O)C=C(C=C1O)C([O-])=O

Specifications

Melting Point 266°C to 207°C
Quantity 1 g
Synonym 3,5-dihydroxy-p-toluic acid, benzoic acid, 3,5-dihydroxy-4-methyl, 4-methyl-3,5-dihydroxy benzoic acid, 3,5-dihydroxy-4-methyl-benzoic acid, pubchem3868, acmc-20a50j, paragos 440034, 3,5-dihydroxy4-methylbenzoic acid, 3,5-dihydroxy-4-methyl benzoic acid, benzoic acid,3,5-dihydroxy-4-methyl
Formula Weight 168.15
Percent Purity 97%
Chemical Name or Material 3,5-Dihydroxy-4-methylbenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only