missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluoropyridine 98.0+%, TCI America™

Catalog No. D40381G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40381G 1 g
D40385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D40381G Supplier TCI America Supplier No. D40381G

Chemical Identifiers

CAS 71902-33-5
Molecular Formula C5H3F2N
Molecular Weight (g/mol) 115.083
MDL Number MFCD03412222
InChI Key WRXAZPPGFLETFR-UHFFFAOYSA-N
Synonym pyridine, 3,5-difluoro, 3,5-difluoro-pyridine, abbypharma ap-14-5380, 3,5-difluoropyridine;, pubchem10556, acmc-209okl, 3,5-difluoro-pyridine;, ksc377a3n, 3,5-difluoropyridine, 3,5-difluoro pyridine
PubChem CID 642820
IUPAC Name 3,5-difluoropyridine
SMILES C1=C(C=NC=C1F)F

Specifications

Color Yellow
Boiling Point 93°C
Quantity 1 g
UN Number 1993
Formula Weight 115.08
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,5-Difluoropyridine