accessibility menu, dialog, popup

UserName

FNF

CAS RN 71902-33-5

IUPAC Name: 3,5-difluoropyridine
Synonyms: 3,5-difluoropyridine
Molecular Weight (g/mol): 115.08
Molecular Formula: C5H3F2N
InChi Key: WRXAZPPGFLETFR-UHFFFAOYSA-N
SMILES: FC1=CC(F)=CN=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)