missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluorophenylmagnesium bromide, 0.50M in 2-MeTHF

Catalog No. AAH54622AE
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH54622AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH54622AE Supplier Thermo Scientific Chemicals Supplier No. H54622AE
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 62351-47-7 | C6H3BrF2Mg | 217.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 62351-47-7
Molecular Formula C6H3BrF2Mg
Molecular Weight (g/mol) 217.30
MDL Number MFCD00672005
InChI Key CURGXPMIRCWEDB-UHFFFAOYSA-M
Synonym 3,5-difluorophenylmagnesium bromide, bromo 3,5-difluorophenyl magnesium, 3,5-difluorophenylmagnesiumbromide, curgxpmircwedb-uhfffaoysa-m, 3,5-difluorophenyl magnesium bromide, 3,5-difluorophenylmagnesium bromide, 0.50m in 2-methf, 3,5-difluorophenylmagnesium bromide 0.5 m in tetrahydrofuran, 3,5-difluorophenylmagnesium bromide solution, 0.5 m in thf, grignard reagent
PubChem CID 2778376
IUPAC Name magnesium;1,3-difluorobenzene-5-ide;bromide
SMILES FC1=CC([Mg]Br)=CC(F)=C1

Specifications

Quantity 100 mL
UN Number UN2924
Sensitivity Air and moisture sensitive; May form a precipitate
Formula Weight 217.29
Concentration or Composition (by Analyte or Components) 0.50M in 2-MeTHF
Chemical Name or Material 3,5-Difluorophenylmagnesium bromide
Hazard Category H225-H302-H314-H335
Hazard Statement GHS H Statement
H225-H314
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only