missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluorophenylhydrazine hydrochloride, 97%

Catalog No. p-7028781
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2618903 1 g
AAH2618906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2618903 Supplier Thermo Scientific Chemicals Supplier No. H2618903
Only null left

CAS: 502496-27-7 | C6H7ClF2N2 | 180.583 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 502496-27-7
Molecular Formula C6H7ClF2N2
Molecular Weight (g/mol) 180.583
MDL Number MFCD03094171
InChI Key CKCNKYAHVKNKHQ-UHFFFAOYSA-N
Synonym 3,5-difluorophenylhydrazine hydrochloride, 3,5-difluorophenyl hydrazine hydrochloride, 3,5-difluorophenylhydrazine hcl, 3,5-difluorophenylhydrazinehydrochloride, 1-3,5-difluorophenyl hydrazine hydrochloride, 3,5-difluoro phenylhydrazine, hydrazine, 3,5-difluorophenyl-, monohydrochloride, pubchem1085, pubchem20776, c6h6f2n2.hcl
PubChem CID 16217172
IUPAC Name (3,5-difluorophenyl)hydrazine;hydrochloride
SMILES C1=C(C=C(C=C1F)F)NN.Cl

Specifications

Melting Point 264°C to 266°C
Quantity 1 g
Formula Weight 180.59
Percent Purity 97%
Chemical Name or Material 3,5-Difluorophenylhydrazine hydrochloride
Hazard Category H301+H311+H331-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only