missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. p-7127604
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D30815G 5 g
D308125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D30815G Supplier TCI America Supplier No. D30815G
Only null left

Chemical Identifiers

CAS 156545-07-2
Molecular Formula C6H5BF2O2
Molecular Weight (g/mol) 157.911
MDL Number MFCD01318138
InChI Key QWQBQRYFWNIDOC-UHFFFAOYSA-N
Synonym 3,5-difluorobenzeneboronic acid, 3,5-difluorophenyl boronic acid, 3,5-difluorophenyl boranediol, 3,5-difluoro phenylboric acid, boronic acid, 3,5-difluorophenyl, 3,5-difluorophenyl dihydroxyborane, 3,5-difluoro phenylboronic acid, boronic acid, b-3,5-difluorophenyl, 3,5-difluorophenylbornic acid
PubChem CID 2734338
IUPAC Name (3,5-difluorophenyl)boronic acid
SMILES B(C1=CC(=CC(=C1)F)F)(O)O

Specifications

Melting Point 250°C
Color White-Yellow
Quantity 5 g
Formula Weight 157.91
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Difluorophenylboronic Acid (contains varying amounts of Anhydride)