missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluorophenylacetic Acid 98.0+%, TCI America™

Catalog No. D34621G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D34621G 1 g
D34625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D34621G Supplier TCI America Supplier No. D34621G
Only null left

Chemical Identifiers

CAS 105184-38-1
Molecular Formula C8H6F2O2
Molecular Weight (g/mol) 172.131
MDL Number MFCD00010316
InChI Key IGGNSAVLXJKCNH-UHFFFAOYSA-N
Synonym 3,5-difluorophenylacetic acid, 2-3,5-difluorophenyl acetic acid, 3,5-difluorophenylaceticacid, 3,5-difluorobenzeneacetic acid, 3,5-difluorophenyl acetic acid, 3,5-difluoro-phenyl-acetic acid, benzeneacetic acid, 3,5-difluoro, 3,5-diifluorophenylacetic acid, 3,5-difluorophenylac
PubChem CID 145424
IUPAC Name 2-(3,5-difluorophenyl)acetic acid
SMILES C1=C(C=C(C=C1F)F)CC(=O)O

Specifications

Melting Point 69°C
Color White-Yellow
Quantity 1 g
Formula Weight 172.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Difluorophenylacetic Acid