missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Difluorobenzylamine 98.0+%, TCI America™
$54.06 - $214.37
Chemical Identifiers
| CAS | 90390-27-5 |
|---|---|
| Molecular Formula | C7H7F2N |
| Molecular Weight (g/mol) | 143.14 |
| MDL Number | MFCD00061244 |
| InChI Key | VJNGGOMRUHYAMC-UHFFFAOYSA-N |
| Synonym | 3,5-difluorobenzylamine, 3,5-difluorophenyl methanamine, benzenemethanamine, 3,5-difluoro, 1-3,5-difluorophenyl methanamine, 3,5-difluorobenzyl amine, 3,5-difluorophenyl methylamine, 3, 5-difluorobenzylamine, pubchem4409, 3,5-difluoro-benzylamine, 3,5-difluoro-benzyl amine |
| PubChem CID | 145211 |
| IUPAC Name | 1-(3,5-difluorophenyl)methanamine |
| SMILES | NCC1=CC(F)=CC(F)=C1 |
Chemical Identifiers
| 90390-27-5 | |
| 143.14 | |
| VJNGGOMRUHYAMC-UHFFFAOYSA-N | |
| 145211 | |
| NCC1=CC(F)=CC(F)=C1 |
| C7H7F2N | |
| MFCD00061244 | |
| 3,5-difluorobenzylamine, 3,5-difluorophenyl methanamine, benzenemethanamine, 3,5-difluoro, 1-3,5-difluorophenyl methanamine, 3,5-difluorobenzyl amine, 3,5-difluorophenyl methylamine, 3, 5-difluorobenzylamine, pubchem4409, 3,5-difluoro-benzylamine, 3,5-difluoro-benzyl amine | |
| 1-(3,5-difluorophenyl)methanamine |
Specifications
| 90390-27-5 | |
| 184°C | |
| MFCD00061244 | |
| 2735 | |
| VJNGGOMRUHYAMC-UHFFFAOYSA-N | |
| 1-(3,5-difluorophenyl)methanamine | |
| 145211 | |
| ≥98.0% (GC,T) | |
| 3,5-Difluorobenzylamine |
| Yellow | |
| C7H7F2N | |
| 1 g | |
| 3,5-difluorobenzylamine, 3,5-difluorophenyl methanamine, benzenemethanamine, 3,5-difluoro, 1-3,5-difluorophenyl methanamine, 3,5-difluorobenzyl amine, 3,5-difluorophenyl methylamine, 3, 5-difluorobenzylamine, pubchem4409, 3,5-difluoro-benzylamine, 3,5-difluoro-benzyl amine | |
| NCC1=CC(F)=CC(F)=C1 | |
| 143.14 | |
| 143.14 | |
| Liquid |
Safety and Handling
TSCA : No