missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluorobenzonitrile 99.0+%, TCI America™

Catalog No. D20815G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D20815G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D20815G Supplier TCI America Supplier No. D20815G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 64248-63-1
Molecular Formula C7H3F2N
Molecular Weight (g/mol) 139.11
MDL Number MFCD00010311
InChI Key CQXZSEXZQVKCHW-UHFFFAOYSA-N
Synonym benzonitrile, 3,5-difluoro, 5-cyano-1,3-difluorobenzene, 3,5-difluoro benzonitrile, 3,5-difluoro-benzonitrile, 3,5-difluorobenzenecarbonitrile, pubchem4761, 3.5-difluorobenzonitrile, acmc-1bc3f, ksc355k7h, paragos 390210
PubChem CID 587204
IUPAC Name 3,5-difluorobenzonitrile
SMILES FC1=CC(=CC(F)=C1)C#N

Specifications

Melting Point 88°C
Color White
Boiling Point 160°C
Quantity 5 g
Formula Weight 139.10
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Difluorobenzonitrile