missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluorobenzonitrile, 99%

CAS: 64248-63-1 | C7H3F2N | 139.11 g/mol

$75.74 - $263.03

Chemical Identifiers

CAS 64248-63-1
Molecular Formula C7H3F2N
Molecular Weight (g/mol) 139.11
MDL Number MFCD00010311
InChI Key CQXZSEXZQVKCHW-UHFFFAOYSA-N
Synonym benzonitrile, 3,5-difluoro, 5-cyano-1,3-difluorobenzene, 3,5-difluoro benzonitrile, 3,5-difluoro-benzonitrile, 3,5-difluorobenzenecarbonitrile, pubchem4761, 3.5-difluorobenzonitrile, acmc-1bc3f, ksc355k7h, paragos 390210
PubChem CID 587204
IUPAC Name 3,5-difluorobenzonitrile
SMILES FC1=CC(=CC(F)=C1)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2134706
View Documents
Thermo Scientific Chemicals
B2134706
5 g
Each for $75.74
Only null left
 
AAB2134714
View Documents
Thermo Scientific Chemicals
B2134714
25 g
Each for $263.03
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

64248-63-1
139.11
CQXZSEXZQVKCHW-UHFFFAOYSA-N
587204
FC1=CC(=CC(F)=C1)C#N
C7H3F2N
MFCD00010311
benzonitrile, 3,5-difluoro, 5-cyano-1,3-difluorobenzene, 3,5-difluoro benzonitrile, 3,5-difluoro-benzonitrile, 3,5-difluorobenzenecarbonitrile, pubchem4761, 3.5-difluorobenzonitrile, acmc-1bc3f, ksc355k7h, paragos 390210
3,5-difluorobenzonitrile

Specifications

64248-63-1
160°C
MFCD00010311
UN3439
benzonitrile, 3,5-difluoro, 5-cyano-1,3-difluorobenzene, 3,5-difluoro benzonitrile, 3,5-difluoro-benzonitrile, 3,5-difluorobenzenecarbonitrile, pubchem4761, 3.5-difluorobenzonitrile, acmc-1bc3f, ksc355k7h, paragos 390210
FC1=CC(=CC(F)=C1)C#N
139.11
139.11
3,5-Difluorobenzonitrile
83°C to 88°C
C7H3F2N
5 g
2082206
CQXZSEXZQVKCHW-UHFFFAOYSA-N
3,5-difluorobenzonitrile
587204
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

EINECSNumber : 264-752-3

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only