missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluorobenzenesulfonamide, 98%

Catalog No. AAL1972006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1972006 5 g
AAL1972003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1972006 Supplier Thermo Scientific Chemicals Supplier No. L1972006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 140480-89-3 | C6H5F2NO2S | 193.168 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 140480-89-3
Molecular Formula C6H5F2NO2S
Molecular Weight (g/mol) 193.168
MDL Number MFCD02091379
InChI Key MKQPOVUFDWKPNO-UHFFFAOYSA-N
Synonym 3,5-difluorobenzenesulphonamide, 3,5-difluorobenzene-1-sulfonamide, benzenesulfonamide, 3,5-difluoro, buttpark 27\07-04, fbu, pubchem11783, acmc-20ai3d, d0s6gk, ksc493s5p, 3,5-difluorobenzenesulfonamide
PubChem CID 446275
ChEBI CHEBI:42566
IUPAC Name 3,5-difluorobenzenesulfonamide
SMILES C1=C(C=C(C=C1F)S(=O)(=O)N)F

Specifications

Melting Point 157°C to 161°C
Quantity 5 g
Beilstein 5333160
Formula Weight 193.17
Percent Purity 98%
Chemical Name or Material 3,5-Difluorobenzenesulfonamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only