missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluorobenzeneboronic acid, 97+%

Catalog No. AAL1742503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1742503 1 g
AAL1742506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1742503 Supplier Thermo Scientific Chemicals Supplier No. L1742503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 156545-07-2 | C6H5BF2O2 | 157.911 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 156545-07-2
Molecular Formula C6H5BF2O2
Molecular Weight (g/mol) 157.911
MDL Number MFCD01318138
InChI Key QWQBQRYFWNIDOC-UHFFFAOYSA-N
Synonym 3,5-difluorobenzeneboronic acid, 3,5-difluorophenyl boronic acid, 3,5-difluorophenyl boranediol, 3,5-difluoro phenylboric acid, boronic acid, 3,5-difluorophenyl, 3,5-difluorophenyl dihydroxyborane, 3,5-difluoro phenylboronic acid, boronic acid, b-3,5-difluorophenyl, 3,5-difluorophenylbornic acid
PubChem CID 2734338
IUPAC Name (3,5-difluorophenyl)boronic acid
SMILES B(C1=CC(=CC(=C1)F)F)(O)O

Specifications

Melting Point >300°C
Quantity 1 g
Beilstein 6800882
Formula Weight 157.91
Percent Purity ≥97%
Chemical Name or Material 3,5-Difluorobenzeneboronic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only