missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluorobenzaldehyde, 97%

Catalog No. AAA1691703
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1691703 1 g
AAA1691706 5 g
AAA1691714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1691703 Supplier Thermo Scientific Chemicals Supplier No. A1691703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 32085-88-4 | C7H4F2O | 142.105 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 32085-88-4
Molecular Formula C7H4F2O
Molecular Weight (g/mol) 142.105
MDL Number MFCD00010329
InChI Key ASOFZHSTJHGQDT-UHFFFAOYSA-N
Synonym benzaldehyde, 3,5-difluoro, 3,5-difluoro-benzaldehyde, pubchem1414, 3,5difluorobenzaldehyde, acmc-1aewe, 3,5-diflourobenzaldehyde, 3-5-difluorobenzaldehyde, 3, 5-difluorobenzaldehyde, ksc224o7p, 3,5-difluorophenyl methanone
PubChem CID 588160
IUPAC Name 3,5-difluorobenzaldehyde
SMILES C1=C(C=C(C=C1F)F)C=O

Specifications

Boiling Point 61°C to 63°C (43 mmHg)
Flash Point 56°C (132°F)
Refractive Index 1.493
Quantity 1 g
UN Number UN1989
Beilstein 2573392
Sensitivity Air Sensitive
Formula Weight 142.11
Percent Purity 97%
Chemical Name or Material 3,5-Difluorobenzaldehyde
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALDEHYDES, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only