missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluoroanisole, 97%, Thermo Scientific Chemicals

Catalog No. p-7022629
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1549714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1549714 Supplier Thermo Scientific Chemicals Supplier No. A1549714

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 93343-10-3
Molecular Formula C7H6F2O
Molecular Weight (g/mol) 144.12
MDL Number MFCD00042560
InChI Key OTGQPYSISUUHAF-UHFFFAOYSA-N
Synonym 3,5-difluoroanisole, 3,5-difluoro-1-methoxybenzene, benzene, 1,3-difluoro-5-methoxy, 1,3-difluoro-5-methoxy-benzene, 3,5-difluoro anisole, 3,5-difluoroanisol, 3.5-difluoroanisole, pubchem1967, 3,5-difluoroanisole;, 3,5,-difluoroanisole;
PubChem CID 2724518
IUPAC Name 1,3-difluoro-5-methoxybenzene
SMILES COC1=CC(F)=CC(F)=C1

Specifications

Density 1.234
Boiling Point 145°C to 148°C
Flash Point 43°C (109°F)
Refractive Index 1.466
Quantity 25 g
UN Number UN1993
Beilstein 4966844
Formula Weight 144.12
Percent Purity 97%
Chemical Name or Material 3,5-Difluoroanisole
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only