missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluoroaniline 98.0+%, TCI America™

Catalog No. p-7127433
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26655G 5 g
D266525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D26655G Supplier TCI America Supplier No. D26655G
Only null left

Chemical Identifiers

CAS 372-39-4
Molecular Formula C6H5F2N
Molecular Weight (g/mol) 129.11
MDL Number MFCD00007763
InChI Key KQOIBXZRCYFZSO-UHFFFAOYSA-N
Synonym benzenamine, 3,5-difluoro, 3,5-difluorobenzenamine, 3,5-difluorophenylamine, 5an, 3,5-difluoranilin, 3,5-difluoroanilin, 3,5 difluoroaniline, 3.5-difluoroaniline, pubchem2296, 3, 5-difluoroaniline
PubChem CID 96595
IUPAC Name 3,5-difluoroaniline
SMILES C1=C(C=C(C=C1F)F)N

Specifications

Melting Point 39°C
Color White-Yellow
Boiling Point 82°C
Quantity 5 g
UN Number 2811
Formula Weight 129.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Difluoroaniline