missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluoroaniline, 98%

Catalog No. AC311420500
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC311420500 50 g Glass bottle
AC311420100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC311420500 Supplier Thermo Scientific Chemicals Supplier No. 311420500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 372-39-4 | C6H5F2N | 129.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 372-39-4
Molecular Formula C6H5F2N
Molecular Weight (g/mol) 129.11
MDL Number MFCD00007763
InChI Key KQOIBXZRCYFZSO-UHFFFAOYSA-N
Synonym benzenamine, 3,5-difluoro, 3,5-difluorobenzenamine, 3,5-difluorophenylamine, 5an, 3,5-difluoranilin, 3,5-difluoroanilin, 3,5 difluoroaniline, 3.5-difluoroaniline, pubchem2296, 3, 5-difluoroaniline
PubChem CID 96595
IUPAC Name 3,5-difluoroaniline
SMILES C1=C(C=C(C=C1F)F)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 37°C to 41°C
Color White to Yellow
Density 75g/mL
Boiling Point 82°C (20.0 mmHg)
Flash Point 75°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula F2C6H3NH2
Quantity 50 g
Formula Weight 129.11
Percent Purity 98%
Physical Form Low Melting Solid
Chemical Name or Material 3, 5-Difluoroaniline
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 206-752-8
Recommended Storage Product darkens on storage

RUO – Research Use Only