missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluoroaniline, 98+%

Catalog No. AAA1652806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1652806 5 g
AAA1652814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1652806 Supplier Thermo Scientific Chemicals Supplier No. A1652806

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 372-39-4 | C6H5F2N | 129.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 372-39-4
Molecular Formula C6H5F2N
Molecular Weight (g/mol) 129.11
MDL Number MFCD00007763
InChI Key KQOIBXZRCYFZSO-UHFFFAOYSA-N
Synonym benzenamine, 3,5-difluoro, 3,5-difluorobenzenamine, 3,5-difluorophenylamine, 5an, 3,5-difluoranilin, 3,5-difluoroanilin, 3,5 difluoroaniline, 3.5-difluoroaniline, pubchem2296, 3, 5-difluoroaniline
PubChem CID 96595
IUPAC Name 3,5-difluoroaniline
SMILES C1=C(C=C(C=C1F)F)N

Specifications

Melting Point 37°C to 41°C
Density 1.295
Boiling Point 80°C (20 mmHg)
Flash Point 75°C (167°F)
Odor Characteristic
Quantity 5 g
UN Number UN2811
Beilstein 2802286
Formula Weight 129.11
Percent Purity ≥98%
Chemical Name or Material 3,5-Difluoroaniline
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H311-H331-H373-H302-H315-H319
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 206-752-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only