Learn More
3,5-Difluoro-N-Fmoc-L-phenylalanine, 95%
CAS: 205526-24-5 | C24H19F2NO4 | 423.42 g/mol
Supplier: Thermo Scientific Chemicals H5206906
Description
3,5-Difluoro-N-Fmoc-L-phenylalanine is used for producing high quality peptides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,5-Difluoro-N-Fmoc-L-phenylalanine is used for producing high quality peptides.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
| 3,5-Difluoro-N-Fmoc-L-phenylalanine | |
| C24H19F2NO4 | |
| 5 g | |
| Slightly soluble in water. | |
| OC(=O)[C@H](CC1=CC(F)=CC(F)=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 | |
| 423.42 | |
| 423.4 |
| 205526-24-5 | |
| MFCD00797579 | |
| fmoc-phe 3,5-f2-oh, fmoc-phe 3,5-dif-oh, fmoc-3,5-difluoro-l-phenylalanine, s-n-fmoc-3,5-difluorophenylalanine, fmoc-l-3,5-difluorophenylalanine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-3,5-difluorophenyl propanoic acid, fmoc-l-3,5-difluorophe, 3,5-difluoro-l-phenylalanine, n-fmoc protected, 2s-3-3,5-difluorophenyl-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid, 2s-3-3,5-difluorophenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid | |
| UYEQBZISDRNPFC-QFIPXVFZSA-N | |
| (2S)-3-(3,5-difluorophenyl)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid | |
| 2774201 | |
| 95% |
Chemical Identifiers
| 205526-24-5 | |
| 423.42 | |
| UYEQBZISDRNPFC-QFIPXVFZSA-N | |
| 2774201 |
| C24H19F2NO4 | |
| MFCD00797579 | |
| fmoc-phe 3,5-f2-oh, fmoc-phe 3,5-dif-oh, fmoc-3,5-difluoro-l-phenylalanine, s-n-fmoc-3,5-difluorophenylalanine, fmoc-l-3,5-difluorophenylalanine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-3,5-difluorophenyl propanoic acid, fmoc-l-3,5-difluorophe, 3,5-difluoro-l-phenylalanine, n-fmoc protected, 2s-3-3,5-difluorophenyl-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid, 2s-3-3,5-difluorophenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid | |
| OC(=O)[C@H](CC1=CC(F)=CC(F)=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only