missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluoro-N-Fmoc-L-phenylalanine, 95%

Catalog No. p-7034386
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52069MD 250 mg
AAH5206903 1 g
AAH5206906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH52069MD Supplier Thermo Scientific Chemicals Supplier No. H52069MD

CAS: 205526-24-5 | C24H19F2NO4 | 423.42 g/mol

3,5-Difluoro-N-Fmoc-L-phenylalanine is used for producing high quality peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Difluoro-N-Fmoc-L-phenylalanine is used for producing high quality peptides.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 205526-24-5
Molecular Formula C24H19F2NO4
Molecular Weight (g/mol) 423.42
MDL Number MFCD00797579
InChI Key UYEQBZISDRNPFC-QFIPXVFZSA-N
Synonym fmoc-phe 3,5-f2-oh, fmoc-phe 3,5-dif-oh, fmoc-3,5-difluoro-l-phenylalanine, s-n-fmoc-3,5-difluorophenylalanine, fmoc-l-3,5-difluorophenylalanine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-3,5-difluorophenyl propanoic acid, fmoc-l-3,5-difluorophe, 3,5-difluoro-l-phenylalanine, n-fmoc protected, 2s-3-3,5-difluorophenyl-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid, 2s-3-3,5-difluorophenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid
PubChem CID 2774201
SMILES OC(=O)[C@H](CC1=CC(F)=CC(F)=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Quantity 250 mg
Solubility Information Slightly soluble in water.
IUPAC Name (2S)-3-(3,5-difluorophenyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
Formula Weight 423.4
Percent Purity 95%
Chemical Name or Material 3,5-Difluoro-N-Fmoc-L-phenylalanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only