missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluoro-N-Fmoc-L-phenylalanine, 95%

CAS: 205526-24-5 | C24H19F2NO4 | 423.42 g/mol

Supplier:  Thermo Scientific Chemicals H5206903

 View more versions of this product

Catalog No. AAH5206903


Only null left
Explore available promotions

Description

Description

3,5-Difluoro-N-Fmoc-L-phenylalanine is used for producing high quality peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Difluoro-N-Fmoc-L-phenylalanine is used for producing high quality peptides.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

3,5-Difluoro-N-Fmoc-L-phenylalanine
C24H19F2NO4
1 g
Slightly soluble in water.
OC(=O)[C@H](CC1=CC(F)=CC(F)=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
423.42
423.4
205526-24-5
MFCD00797579
fmoc-phe 3,5-f2-oh, fmoc-phe 3,5-dif-oh, fmoc-3,5-difluoro-l-phenylalanine, s-n-fmoc-3,5-difluorophenylalanine, fmoc-l-3,5-difluorophenylalanine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-3,5-difluorophenyl propanoic acid, fmoc-l-3,5-difluorophe, 3,5-difluoro-l-phenylalanine, n-fmoc protected, 2s-3-3,5-difluorophenyl-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid, 2s-3-3,5-difluorophenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid
UYEQBZISDRNPFC-QFIPXVFZSA-N
(2S)-3-(3,5-difluorophenyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
2774201
95%

Chemical Identifiers

205526-24-5
423.42
UYEQBZISDRNPFC-QFIPXVFZSA-N
2774201
C24H19F2NO4
MFCD00797579
fmoc-phe 3,5-f2-oh, fmoc-phe 3,5-dif-oh, fmoc-3,5-difluoro-l-phenylalanine, s-n-fmoc-3,5-difluorophenylalanine, fmoc-l-3,5-difluorophenylalanine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-3,5-difluorophenyl propanoic acid, fmoc-l-3,5-difluorophe, 3,5-difluoro-l-phenylalanine, n-fmoc protected, 2s-3-3,5-difluorophenyl-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid, 2s-3-3,5-difluorophenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid
OC(=O)[C@H](CC1=CC(F)=CC(F)=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only