missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluoro-4-hydroxybenzonitrile, 97%

Catalog No. AAH31620MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH31620MD 250 mg
AAH3162003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH31620MD Supplier Thermo Scientific Chemicals Supplier No. H31620MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2967-54-6 | C7H3F2NO | 155.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2967-54-6
Molecular Formula C7H3F2NO
Molecular Weight (g/mol) 155.10
MDL Number MFCD09033181
InChI Key XZNZJDPPWWFJAL-UHFFFAOYSA-N
Synonym 3,5-difluoro-4-hydroxy-benzonitrile, 2,6-difluoro-4-cyanophenol, 4-cyano-2,6-difluorophenol, 3,5-difluoro-4-hydroxy benzonitrile, pubchem23504, benzonitrile, 3,5-difluoro-4-hydroxy
PubChem CID 10749406
IUPAC Name 3,5-difluoro-4-hydroxybenzonitrile
SMILES OC1=C(F)C=C(C=C1F)C#N

Specifications

Melting Point 200°C to 208°C
Quantity 250 mg
UN Number UN3439
Formula Weight 155.1
Percent Purity 97%
Chemical Name or Material 3,5-Difluoro-4-hydroxybenzonitrile
Hazard Category H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only