missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluoro-4-hydroxybenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Catalog No. p-7054121
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH62042MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH62042MD Supplier Thermo Scientific Chemicals Supplier No. H62042MD

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1029439-83-5
Molecular Formula C12H15BF2O3
Molecular Weight (g/mol) 256.056
MDL Number MFCD16996286
InChI Key KTQYJNVSVKKLGQ-UHFFFAOYSA-N
Synonym 2,6-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 3,5-difluoro-4-hydroxybenzeneboronic acid pinacol ester, 3,5-difluoro-4-hydroxyphenylboronic acid pinacol ester
PubChem CID 57655406
IUPAC Name 2,6-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C(=C2)F)O)F

Specifications

Color White
Quantity 250 mg
Formula Weight 256.05
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 3,5-Difluoro-4-hydroxybenzeneboronic acid pinacol ester
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only