missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorosulfanilamide 98.0+%, TCI America™

$78.93 - $242.57

Chemical Identifiers

CAS 22134-75-4
Molecular Formula C6H6Cl2N2O2S
Molecular Weight (g/mol) 241.086
MDL Number MFCD00014784
InChI Key DVZMRTJKNJKEGV-UHFFFAOYSA-N
Synonym 3,5-dichlorosulfanilamide, benzenesulfonamide, 4-amino-3,5-dichloro, 4-amino-3,5-dichlorobenzene-1-sulfonamide, sulfanilamide, 3,5-dichloro, 4-amino-3,5-dichloro-benzenesulfonamide, 3,5-dichlorosulphanilamide, 1zge, acmc-1clhm, sulfanilamide,5-dichloro, aronis24244
PubChem CID 89607
IUPAC Name 4-amino-3,5-dichlorobenzenesulfonamide
SMILES C1=C(C=C(C(=C1Cl)N)Cl)S(=O)(=O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D04145G
View Documents
TCI America
D04145G
5 g
Each for $78.93
Only null left
 
D041425G
View Documents
TCI America
D041425G
25 g
Each for $242.57
Only null left
 
Description

Description

Specifications

Chemical Identifiers

22134-75-4
241.086
DVZMRTJKNJKEGV-UHFFFAOYSA-N
89607
C1=C(C=C(C(=C1Cl)N)Cl)S(=O)(=O)N
C6H6Cl2N2O2S
MFCD00014784
3,5-dichlorosulfanilamide, benzenesulfonamide, 4-amino-3,5-dichloro, 4-amino-3,5-dichlorobenzene-1-sulfonamide, sulfanilamide, 3,5-dichloro, 4-amino-3,5-dichloro-benzenesulfonamide, 3,5-dichlorosulphanilamide, 1zge, acmc-1clhm, sulfanilamide,5-dichloro, aronis24244
4-amino-3,5-dichlorobenzenesulfonamide

Specifications

22134-75-4
C6H6Cl2N2O2S
5 g
DVZMRTJKNJKEGV-UHFFFAOYSA-N
4-amino-3,5-dichlorobenzenesulfonamide
89607
≥98.0% (N)
3,5-Dichlorosulfanilamide
206°C
MFCD00014784
3,5-dichlorosulfanilamide, benzenesulfonamide, 4-amino-3,5-dichloro, 4-amino-3,5-dichlorobenzene-1-sulfonamide, sulfanilamide, 3,5-dichloro, 4-amino-3,5-dichloro-benzenesulfonamide, 3,5-dichlorosulphanilamide, 1zge, acmc-1clhm, sulfanilamide,5-dichloro, aronis24244
C1=C(C=C(C(=C1Cl)N)Cl)S(=O)(=O)N
241.086
241.09
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents