missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorophenylhydrazine Hydrochloride 90.0+%, TCI America™

Catalog No. D26445G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26445G 5 g
D264425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D26445G Supplier TCI America Supplier No. D26445G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 63352-99-8
Molecular Formula C6H7Cl3N2
Molecular Weight (g/mol) 213.486
MDL Number MFCD00012938
InChI Key BDXZOJVMTJOAPS-UHFFFAOYSA-N
Synonym 3,5-dichlorophenylhydrazine hydrochloride, 3,5-dichlorophenyl hydrazine hydrochloride, 3,5-dichlorophenyl hydrazine monohydrochloride, 3,5-dichlorophenylhydrazine hcl, 1-3,5-dichlorophenyl hydrazine hydrochloride, hydrazine, 3,5-dichlorophenyl-, monohydrochloride, 3,5-dichlorophenylhydrazinehydrochloride, pubchem7531, c6h6cl2n2.hcl, acmc-209nf3
PubChem CID 2723913
IUPAC Name (3,5-dichlorophenyl)hydrazine;hydrochloride
SMILES C1=C(C=C(C=C1Cl)Cl)NN.Cl

Specifications

Quantity 5 g
Formula Weight 213.49
Percent Purity ≥90.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dichlorophenylhydrazine Hydrochloride
EINECSNumber (3)-0472
TSCA No