Learn More
3,5-Dichlorophenylacetic acid, 95%
CAS: 51719-65-4 | C8H6Cl2O2 | 205.034 g/mol
$122.16 - $467.81
Chemical Identifiers
| CAS | 51719-65-4 |
|---|---|
| Molecular Formula | C8H6Cl2O2 |
| Molecular Weight (g/mol) | 205.034 |
| MDL Number | MFCD01764671 |
| InChI Key | RERINLRFXYGZEE-UHFFFAOYSA-N |
| Synonym | 2-3,5-dichlorophenyl acetic acid, 3,5-dichlorophenylacetic acid, benzeneacetic acid, 3,5-dichloro, 3,5-dichlorobenzeneacetic acid, 3,5-dichlorophenyl acetic acid, 3,5-dichloro-phenyl-acetic acid, 2-3,5-dichlorophenyl-acetic acid, 3,5-dichlorophenylaceticacid, pubchem24116, 3,5-dichlorphenylacetic acid |
| PubChem CID | 6452487 |
| IUPAC Name | 2-(3,5-dichlorophenyl)acetic acid |
| SMILES | C1=C(C=C(C=C1Cl)Cl)CC(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6329303
|
Thermo Scientific Chemicals
H6329303 |
1 g |
Each for $122.16
|
|
|||||
|
AAH6329306
|
Thermo Scientific Chemicals
H6329306 |
5 g |
Each for $467.81
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 51719-65-4 | |
| 205.034 | |
| RERINLRFXYGZEE-UHFFFAOYSA-N | |
| 6452487 | |
| C1=C(C=C(C=C1Cl)Cl)CC(=O)O |
| C8H6Cl2O2 | |
| MFCD01764671 | |
| 2-3,5-dichlorophenyl acetic acid, 3,5-dichlorophenylacetic acid, benzeneacetic acid, 3,5-dichloro, 3,5-dichlorobenzeneacetic acid, 3,5-dichlorophenyl acetic acid, 3,5-dichloro-phenyl-acetic acid, 2-3,5-dichlorophenyl-acetic acid, 3,5-dichlorophenylaceticacid, pubchem24116, 3,5-dichlorphenylacetic acid | |
| 2-(3,5-dichlorophenyl)acetic acid |
Specifications
| 51719-65-4 | |
| MFCD01764671 | |
| 2-3,5-dichlorophenyl acetic acid, 3,5-dichlorophenylacetic acid, benzeneacetic acid, 3,5-dichloro, 3,5-dichlorobenzeneacetic acid, 3,5-dichlorophenyl acetic acid, 3,5-dichloro-phenyl-acetic acid, 2-3,5-dichlorophenyl-acetic acid, 3,5-dichlorophenylaceticacid, pubchem24116, 3,5-dichlorphenylacetic acid | |
| C1=C(C=C(C=C1Cl)Cl)CC(=O)O | |
| 205.034 | |
| 205.03 | |
| Solid |
| C8H6Cl2O2 | |
| 1 g | |
| RERINLRFXYGZEE-UHFFFAOYSA-N | |
| 2-(3,5-dichlorophenyl)acetic acid | |
| 6452487 | |
| 95% | |
| 3,5-Dichlorophenylacetic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only