missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorophenylacetic acid, 95%

Catalog No. AAH6329303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6329303 1 g
AAH6329306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6329303 Supplier Thermo Scientific Chemicals Supplier No. H6329303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 51719-65-4 | C8H6Cl2O2 | 205.034 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51719-65-4
Molecular Formula C8H6Cl2O2
Molecular Weight (g/mol) 205.034
MDL Number MFCD01764671
InChI Key RERINLRFXYGZEE-UHFFFAOYSA-N
Synonym 2-3,5-dichlorophenyl acetic acid, 3,5-dichlorophenylacetic acid, benzeneacetic acid, 3,5-dichloro, 3,5-dichlorobenzeneacetic acid, 3,5-dichlorophenyl acetic acid, 3,5-dichloro-phenyl-acetic acid, 2-3,5-dichlorophenyl-acetic acid, 3,5-dichlorophenylaceticacid, pubchem24116, 3,5-dichlorphenylacetic acid
PubChem CID 6452487
IUPAC Name 2-(3,5-dichlorophenyl)acetic acid
SMILES C1=C(C=C(C=C1Cl)Cl)CC(=O)O

Specifications

Quantity 1 g
Formula Weight 205.03
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3,5-Dichlorophenylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only