missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorophenol, 99%

Catalog No. p-3736189
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC113610100 10 g Glass bottle
AC113610500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC113610100 Supplier Thermo Scientific Chemicals Supplier No. 113610100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 591-35-5 | C6H4Cl2O | 163 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 591-35-5
Molecular Formula C6H4Cl2O
Molecular Weight (g/mol) 163
MDL Number MFCD00002259
InChI Key VPOMSPZBQMDLTM-UHFFFAOYSA-N
Synonym phenol, 3,5-dichloro, 3,5-dichloro-phenol, unii-fg32l88ko9, 3,5 dichlorophenol, ccris 5905, 3,5-dichloro phenol, pubchem3699, acmc-209mar, dsstox_cid_5006, dsstox_rid_77624
PubChem CID 11571
IUPAC Name 3,5-dichlorophenol
SMILES C1=C(C=C(C=C1Cl)Cl)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 64°C to 68°C
Color Yellow to Brown
Boiling Point 233°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula Cl2C6H3OH
Quantity 10 g
Beilstein 06, 190
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol,acetone,dmso
Formula Weight 163
Percent Purity 99%
Physical Form Crystalline Powder, Crystals or Crystalline Mass
Chemical Name or Material 3, 5-Dichlorophenol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 209-714-9

RUO – Research Use Only