missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorobenzoyl Chloride 95.0+%, TCI America™

Catalog No. D140425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D140425G 25 g
D1404500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D140425G Supplier TCI America Supplier No. D140425G

Chemical Identifiers

CAS 2905-62-6
Molecular Formula C7H3Cl3O
Molecular Weight (g/mol) 209.45
MDL Number MFCD00009817
InChI Key GGHLXLVPNZMBQR-UHFFFAOYSA-N
Synonym benzoyl chloride, 3,5-dichloro, 3,5-dichlorobenzoylchloride, dichlorobenzoylchloride 3,5-, pubchem10805, dsstox_cid_7506, acmc-209h6l, dsstox_rid_78478, dsstox_gsid_27506, ksc204k6p, 3,5-dichloro benzoyl chloride
PubChem CID 76191
IUPAC Name 3,5-dichlorobenzoyl chloride
SMILES ClC(=O)C1=CC(Cl)=CC(Cl)=C1

Specifications

Color White
Quantity 25 g
UN Number 3261
Formula Weight 209.45
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 3,5-Dichlorobenzoyl Chloride
EINECSNumber (3)-0062&(3)-1409
RTECSNumber DM6636766
TSCA Yes