missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorobenzoic Acid 98.0+%, TCI America™

Catalog No. p-7126158
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D034125G 25 g
D0341500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D034125G Supplier TCI America Supplier No. D034125G
Only null left

Chemical Identifiers

CAS 51-36-5
Molecular Formula C7H4Cl2O2
Molecular Weight (g/mol) 191.007
MDL Number MFCD00002494
InChI Key CXKCZFDUOYMOOP-UHFFFAOYSA-N
Synonym benzoic acid, 3,5-dichloro, 3,5-dichloro-benzoic acid, unii-7baa7r70gn, 7baa7r70gn, ccris 8611, acmc-209ktc, 3,5-dichlorobenzoicacid, 3,5-dichlorobenzic acid, 3 5-dichlorobenzoic acid, dsstox_cid_4981
PubChem CID 5811
IUPAC Name 3,5-dichlorobenzoic acid
SMILES C1=C(C=C(C=C1Cl)Cl)C(=O)O

Specifications

Melting Point 187°C
Color White
Quantity 25 g
Formula Weight 191.01
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dichlorobenzoic Acid
EINECSNumber (3)-1410&(3)-1424
RTECSNumber DG7350000
TSCA No