missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorobenzoic acid, 99%

Catalog No. AAA1516822
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1516822 100 g
AAA1516806 5 g
AAA1516814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1516822 Supplier Thermo Scientific Chemicals Supplier No. A1516822
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 51-36-5 | C7H4Cl2O2 | 191.007 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51-36-5
Molecular Formula C7H4Cl2O2
Molecular Weight (g/mol) 191.007
MDL Number MFCD00002494
InChI Key CXKCZFDUOYMOOP-UHFFFAOYSA-N
Synonym benzoic acid, 3,5-dichloro, 3,5-dichloro-benzoic acid, unii-7baa7r70gn, 7baa7r70gn, ccris 8611, acmc-209ktc, 3,5-dichlorobenzoicacid, 3,5-dichlorobenzic acid, 3 5-dichlorobenzoic acid, dsstox_cid_4981
PubChem CID 5811
IUPAC Name 3,5-dichlorobenzoic acid
SMILES C1=C(C=C(C=C1Cl)Cl)C(=O)O

Specifications

Melting Point 185°C to 187°C
Quantity 100 g
Beilstein 2044776
Formula Weight 191.01
Percent Purity 99%
Chemical Name or Material 3,5-Dichlorobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 200-092-4
RTECSNumber DG7350000
TSCA Yes

RUO – Research Use Only