missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorobenzhydrazide, 97%

Catalog No. AAB2238303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2238303 1 g
AAB2238306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2238303 Supplier Thermo Scientific Chemicals Supplier No. B2238303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 62899-78-9 | C7H6Cl2N2O | 205.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 62899-78-9
Molecular Formula C7H6Cl2N2O
Molecular Weight (g/mol) 205.04
MDL Number MFCD00833411
InChI Key DNISXKBKTSHNKL-UHFFFAOYSA-N
Synonym 3,5-dichlorobenzhydrazide, 3,5-dichlorobenzenecarbohydrazide, acmc-1b4zh, 3,5-bis chloranyl benzohydrazide, 3,5-dichlorobenzoic acid hydrazide, benzoic acid,3,5-dichloro-,hydrazide, benzoic acid,3,5-dichloro-, hydrazide
PubChem CID 2735966
IUPAC Name 3,5-dichlorobenzohydrazide
SMILES NNC(=O)C1=CC(Cl)=CC(Cl)=C1

Specifications

Melting Point 205°C to 207°C
Quantity 1 g
Formula Weight 205.04
Percent Purity 97%
Chemical Name or Material 3,5-Dichlorobenzhydrazide
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only