missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorobenzenethiol 97.0+%, TCI America™

$143.54 - $576.00

Chemical Identifiers

CAS 17231-94-6
Molecular Formula C6H4Cl2S
Molecular Weight (g/mol) 179.058
MDL Number MFCD00041423
InChI Key WRXIPCQPHZMXOO-UHFFFAOYSA-N
Synonym 3,5-dichlorothiophenol, 3,5-dichloro thiophenol, 3,5-dichlorobenzenthiol, 3,5-dichlorobenzene-1-thiol, pubchem6804, 3,5-dichloro-thiophenol, 3,5-dichloro-benzenethiol, acmc-209e4e, 3,5-dichlorophenyl mercaptan, benzenethiol, 3,5-dichloro
PubChem CID 2736096
IUPAC Name 3,5-dichlorobenzenethiol
SMILES C1=C(C=C(C=C1Cl)Cl)S
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D19055G
View Documents
TCI America
D19055G
5 g
Each for $143.54
Only null left
 
D190525G
View Documents
TCI America
D190525G
25 g
Each for $576.00
Only null left
 
Description

Description

Specifications

Chemical Identifiers

17231-94-6
179.058
WRXIPCQPHZMXOO-UHFFFAOYSA-N
2736096
C1=C(C=C(C=C1Cl)Cl)S
C6H4Cl2S
MFCD00041423
3,5-dichlorothiophenol, 3,5-dichloro thiophenol, 3,5-dichlorobenzenthiol, 3,5-dichlorobenzene-1-thiol, pubchem6804, 3,5-dichloro-thiophenol, 3,5-dichloro-benzenethiol, acmc-209e4e, 3,5-dichlorophenyl mercaptan, benzenethiol, 3,5-dichloro
3,5-dichlorobenzenethiol

Specifications

17231-94-6
White-Yellow
C6H4Cl2S
5 g
WRXIPCQPHZMXOO-UHFFFAOYSA-N
3,5-dichlorobenzenethiol
2736096
≥97.0% (GC)
3,5-Dichlorobenzenethiol
63°C
114°C
MFCD00041423
3,5-dichlorothiophenol, 3,5-dichloro thiophenol, 3,5-dichlorobenzenthiol, 3,5-dichlorobenzene-1-thiol, pubchem6804, 3,5-dichloro-thiophenol, 3,5-dichloro-benzenethiol, acmc-209e4e, 3,5-dichlorophenyl mercaptan, benzenethiol, 3,5-dichloro
C1=C(C=C(C=C1Cl)Cl)S
179.058
179.06
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents