missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorobenzenethiol 97.0+%, TCI America™

Catalog No. D19055G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D19055G 5 g
D190525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D19055G Supplier TCI America Supplier No. D19055G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17231-94-6
Molecular Formula C6H4Cl2S
Molecular Weight (g/mol) 179.058
MDL Number MFCD00041423
InChI Key WRXIPCQPHZMXOO-UHFFFAOYSA-N
Synonym 3,5-dichlorothiophenol, 3,5-dichloro thiophenol, 3,5-dichlorobenzenthiol, 3,5-dichlorobenzene-1-thiol, pubchem6804, 3,5-dichloro-thiophenol, 3,5-dichloro-benzenethiol, acmc-209e4e, 3,5-dichlorophenyl mercaptan, benzenethiol, 3,5-dichloro
PubChem CID 2736096
IUPAC Name 3,5-dichlorobenzenethiol
SMILES C1=C(C=C(C=C1Cl)Cl)S

Specifications

Melting Point 63°C
Color White-Yellow
Boiling Point 114°C
Quantity 5 g
Formula Weight 179.06
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dichlorobenzenethiol