missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichloro-4-hydroxybenzoic Acid 98.0+%, TCI America™

Catalog No. p-7126172
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D03735G 5 g
D037325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D03735G Supplier TCI America Supplier No. D03735G
Only null left

Chemical Identifiers

CAS 3336-41-2
Molecular Formula C7H4Cl2O3
Molecular Weight (g/mol) 207.01
MDL Number MFCD00002550
InChI Key AULKDLUOQCUNOK-UHFFFAOYSA-N
Synonym diclhbz, benzoic acid, 3,5-dichloro-4-hydroxy, 3,5-dichloro-4-hydroxy-benzoic acid, pubchem3689, acmc-1cjos, epitope id:122960, 3,5-dichloro-p-salicylic acid, rarechem al be 0933, timtec-bb sbb042792, benzoic acid,5-dichloro-4-hydroxy
PubChem CID 18749
ChEBI CHEBI:53685
IUPAC Name 3,5-dichloro-4-hydroxybenzoic acid
SMILES OC(=O)C1=CC(Cl)=C(O)C(Cl)=C1

Specifications

Color White
Quantity 5 g
Formula Weight 207.01
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dichloro-4-hydroxybenzoic Acid
RTECSNumber DG7502000
TSCA No