missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichloro-4-hydroxybenzoic acid, 97%

CAS: 3336-41-2 | C7H4Cl2O3 | 207.01 g/mol

Supplier:  Thermo Scientific Chemicals A1292322

 View more versions of this product

Catalog No. AAA1292322

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3,5-Dichloro-4-hydroxybenzoic acid
265°C to 270°C
MFCD00002550
2616297
AULKDLUOQCUNOK-UHFFFAOYSA-N
3,5-dichloro-4-hydroxybenzoic acid
18749
207.01
3336-41-2
C7H4Cl2O3
100 g
diclhbz, benzoic acid, 3,5-dichloro-4-hydroxy, 3,5-dichloro-4-hydroxy-benzoic acid, pubchem3689, acmc-1cjos, epitope id:122960, 3,5-dichloro-p-salicylic acid, rarechem al be 0933, timtec-bb sbb042792, benzoic acid,5-dichloro-4-hydroxy
OC(=O)C1=CC(Cl)=C(O)C(Cl)=C1
207.01
CHEBI:53685
97%

Chemical Identifiers

3336-41-2
207.01
AULKDLUOQCUNOK-UHFFFAOYSA-N
18749
3,5-dichloro-4-hydroxybenzoic acid
C7H4Cl2O3
MFCD00002550
diclhbz, benzoic acid, 3,5-dichloro-4-hydroxy, 3,5-dichloro-4-hydroxy-benzoic acid, pubchem3689, acmc-1cjos, epitope id:122960, 3,5-dichloro-p-salicylic acid, rarechem al be 0933, timtec-bb sbb042792, benzoic acid,5-dichloro-4-hydroxy
CHEBI:53685
OC(=O)C1=CC(Cl)=C(O)C(Cl)=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 222-071-9

RTECSNumber : DG7502000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only