missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichloro-4-hydroxybenzoic acid, 97%

Catalog No. AAA1292306
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1292306 5 g
AAA1292314 25 g
AAA1292322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1292306 Supplier Thermo Scientific Chemicals Supplier No. A1292306

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3336-41-2 | C7H4Cl2O3 | 207.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3336-41-2
Molecular Formula C7H4Cl2O3
Molecular Weight (g/mol) 207.01
MDL Number MFCD00002550
InChI Key AULKDLUOQCUNOK-UHFFFAOYSA-N
Synonym diclhbz, benzoic acid, 3,5-dichloro-4-hydroxy, 3,5-dichloro-4-hydroxy-benzoic acid, pubchem3689, acmc-1cjos, epitope id:122960, 3,5-dichloro-p-salicylic acid, rarechem al be 0933, timtec-bb sbb042792, benzoic acid,5-dichloro-4-hydroxy
PubChem CID 18749
ChEBI CHEBI:53685
IUPAC Name 3,5-dichloro-4-hydroxybenzoic acid
SMILES OC(=O)C1=CC(Cl)=C(O)C(Cl)=C1

Specifications

Melting Point 265°C to 270°C
Quantity 5 g
Beilstein 2616297
Formula Weight 207.01
Percent Purity 97%
Chemical Name or Material 3,5-Dichloro-4-hydroxybenzoic acid
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 222-071-9
RTECSNumber DG7502000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only