missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichloro-4-fluoro-1-nitrobenzene, 95%

Catalog No. p-7053704
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6132106 5 g
AAH6132114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6132106 Supplier Thermo Scientific Chemicals Supplier No. H6132106

CAS: 3107-19-5 | C6H2Cl2FNO2 | 209.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3107-19-5
Molecular Formula C6H2Cl2FNO2
Molecular Weight (g/mol) 209.99
MDL Number MFCD00129963
InChI Key VMAATSFMXSMKPG-UHFFFAOYSA-N
Synonym 3,5-dichloro-4-fluoronitrobenzene, benzene, 1,3-dichloro-2-fluoro-5-nitro, 2,6-dichloro-4-nitrofluorobenzene, zlchem 371, pubchem4387, acmc-209hk6, ksc495g6p, 3,5-dichloro-4-fluoro-1-nitrobenzene, 1,3-dichloro-2-fluoro-5-nitro-benzene
PubChem CID 76554
IUPAC Name 1,3-dichloro-2-fluoro-5-nitrobenzene
SMILES [O-][N+](=O)C1=CC(Cl)=C(F)C(Cl)=C1

Specifications

Color White
Quantity 5 g
Formula Weight 209.99
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 3,5-Dichloro-4-fluoro-1-nitrobenzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 221-466-3
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only