missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichloro-2-hydroxybenzenesulfonyl chloride, 97%

Catalog No. AAL1214718
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1214718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1214718 Supplier Thermo Scientific Chemicals Supplier No. L1214718
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 23378-88-3 | C6H3Cl3O3S | 261.50 g/mol

3,5-Dichloro-2-hydroxybenzenesulfonyl chloride is used in the synthesis of (1R,2R)-(+)-1,2-(3,3?,5,5?-tetrachloro-2,2?-dihydroxydibenzenesulfonamido)-1,2-diphenylethane1 and (1R,2R)-(+)- 1,2-(3,3?,5,5?-tetrachloro-2,2?-dihydroxydibenzenesulfonamido)cyclohexane. It was used as chromogenic system in one-step kinetic method for the determination of 5?-nucleotidase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dichloro-2-hydroxybenzenesulfonyl chloride is used in the synthesis of (1R,2R)-(+)-1,2-(3,3′,5,5′-tetrachloro-2,2′-dihydroxydibenzenesulfonamido)-1,2-diphenylethane1 and (1R,2R)-(+)- 1,2-(3,3′,5,5′-tetrachloro-2,2′-dihydroxydibenzenesulfonamido)cyclohexane. It was used as chromogenic system in one-step kinetic method for the determination of 5′-nucleotidase.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Incompatible with bases, moisture and oxidizing agents. Store under dry inert gas. Protect from humidity and water.

Chemical Identifiers

CAS 23378-88-3
Molecular Formula C6H3Cl3O3S
Molecular Weight (g/mol) 261.50
MDL Number MFCD00007432
InChI Key KXFQRJNVGBIDHA-UHFFFAOYSA-N
Synonym 3,5-dichloro-2-hydroxybenzenesulphonyl chloride, benzenesulfonyl chloride, 3,5-dichloro-2-hydroxy, 3,5-dichloro-2-hydroxybenzenesulfonylchloride, 3,5-dichloro-2-hydroxybenzene-1-sulfonyl chloride, 3,5-dichloro-2-hydroxy-benzenesulfonyl chloride, acmc-20andd, kxfqrjnvgbidha-uhfffaoysa, 2,4-dichloro-6-chlorosulfonyl phenol, 2,4-dichlorophenol-6-sulfonyl chloride, 3,5-dichloro-2-hydroxyphenyl chlorosulfone
PubChem CID 90078
IUPAC Name 3,5-dichloro-2-hydroxybenzenesulfonyl chloride
SMILES OC1=C(Cl)C=C(Cl)C=C1S(Cl)(=O)=O

Specifications

Melting Point 78°C to 80°C
Quantity 50 g
UN Number UN3261
Beilstein 2940993
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water.
Formula Weight 261.51
Percent Purity 97%
Chemical Name or Material 3,5-Dichloro-2-hydroxybenzenesulfonyl chloride
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 245-619-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only