missing translation for 'onlineSavingsMsg'
Learn More

3',5'-Dichloro-2,2,2-trifluoroacetophenone 96.0+%, TCI America™

Catalog No. D51135G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D51135G 5 g
D51131G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D51135G Supplier TCI America Supplier No. D51135G
Only null left

Chemical Identifiers

CAS 130336-16-2
Molecular Formula C8H3Cl2F3O
Molecular Weight (g/mol) 243.006
MDL Number MFCD01319996
InChI Key DZDSQRPDUCSOQV-UHFFFAOYSA-N
PubChem CID 2758231
IUPAC Name 1-(3,5-dichlorophenyl)-2,2,2-trifluoroethanone
SMILES C1=C(C=C(C=C1Cl)Cl)C(=O)C(F)(F)F

Specifications

Boiling Point 87°C
Quantity 5 g
Formula Weight 243.01
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 3′,5′-Dichloro-2,2,2-trifluoroacetophenone