accessibility menu, dialog, popup

UserName

CAS RN 130336-16-2

ClClOFFF

CAS RN 130336-16-2

IUPAC Name: 1-(3,5-dichlorophenyl)-2,2,2-trifluoroethan-1-one
Synonyms: 1-(3,5-dichlorophenyl)-2,2,2-trifluoroethanone
Molecular Weight (g/mol): 243.01
Molecular Formula: C8H3Cl2F3O
InChi Key: DZDSQRPDUCSOQV-UHFFFAOYSA-N
SMILES: FC(F)(F)C(=O)C1=CC(Cl)=CC(Cl)=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results