missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromopyridine, 98+%

CAS: 625-92-3 | C5H3Br2N | 236.89 g/mol

Supplier:  Thermo Scientific Chemicals A1087514

 View more versions of this product

Catalog No. AAA1087514


Only null left
Explore available promotions

Description

Description

3-Acetylamino-5-ethoxypyridine was prepared by converting 3, 5-dibromopyridine with sodium ethylate into 3-bromo-5-ethoxypyridine, allowing this substance to interact with ammonia and acetylating the aminoethoxypyridine thus formed with acetic anhydride. Lithiation of 3, 5-dibromopyridine with LDA and subsequent reaction with electrophiles provided 4-alkyl-3, 5-dibromopyridines 2 in high yield. Ligand was prepared by a Pd(0)-catalyzed cross-coupling reaction of 3,5-dibromopyridine and 5-tributylstannyl-3,3?-bipyridine. The synthesis of 3, 5-bis (2-indolyl) pyridine and 3-[(2-indolyl)-5-phenyl] pyridine derivatives as includes Stille or Suzuki type reactions, which were realized on the 3,5-dibromopyridine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Acetylamino-5-ethoxypyridine was prepared by converting 3, 5-dibromopyridine with sodium ethylate into 3-bromo-5-ethoxypyridine, allowing this substance to interact with ammonia and acetylating the aminoethoxypyridine thus formed with acetic anhydride. Lithiation of 3, 5-dibromopyridine with LDA and subsequent reaction with electrophiles provided 4-alkyl-3, 5-dibromopyridines 2 in high yield. Ligand was prepared by a Pd(0)-catalyzed cross-coupling reaction of 3,5-dibromopyridine and 5-tributylstannyl-3,3′-bipyridine. The synthesis of 3, 5-bis (2-indolyl) pyridine and 3-[(2-indolyl)-5-phenyl] pyridine derivatives as includes Stille or Suzuki type reactions, which were realized on the 3,5-dibromopyridine.

Solubility
Soluble in Chloroform and Methanol. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, alkali metals, moisture.
Specifications

Specifications

3,5-Dibromopyridine
110°C to 113°C
MFCD00014634
108477
Soluble in Chloroform and Methanol. Insoluble in water.
BrC1=CC(Br)=CN=C1
236.89
CHEBI:51593
≥98%
625-92-3
C5H3Br2N
25 g
pyridine, 3,5-dibromo, 3,5-dibromo-pyridine, 3,5-dibromo pyridine, 3,5-dibrompyridin, zlchem 300, 3.5-dibromopyridine, 3,5-dibromo-pyridin, pubchem2133, 3, 5-dibromopyridine, acmc-1auln
SOSPMXMEOFGPIM-UHFFFAOYSA-N
3,5-dibromopyridine
69369
236.9

Chemical Identifiers

625-92-3
236.89
SOSPMXMEOFGPIM-UHFFFAOYSA-N
69369
3,5-dibromopyridine
C5H3Br2N
MFCD00014634
pyridine, 3,5-dibromo, 3,5-dibromo-pyridine, 3,5-dibromo pyridine, 3,5-dibrompyridin, zlchem 300, 3.5-dibromopyridine, 3,5-dibromo-pyridin, pubchem2133, 3, 5-dibromopyridine, acmc-1auln
CHEBI:51593
BrC1=CC(Br)=CN=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 210-916-4

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only