missing translation for 'onlineSavingsMsg'
Learn More

(3,5-Dibromophenyl)triphenylsilane 98.0+%, TCI America™

Catalog No. p-7128739
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5338200MG 200 mg
D53381G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D5338200MG Supplier TCI America Supplier No. D5338200MG
Only null left

Chemical Identifiers

CAS 1030856-97-3
Molecular Formula C24H18Br2Si
Molecular Weight (g/mol) 494.301
InChI Key UYGFDYMFXPXXLL-UHFFFAOYSA-N
Synonym 3,5-Dibromotetraphenylsilane
PubChem CID 58926930
IUPAC Name (3,5-dibromophenyl)-triphenylsilane
SMILES C1=CC=C(C=C1)[Si](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC(=CC(=C4)Br)Br

Specifications

Melting Point 188°C
Color White-Yellow
Quantity 200 mg
Formula Weight 494.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (3,5-Dibromophenyl)triphenylsilane