missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromophenol 98.0+%, TCI America™

Catalog No. p-7127936
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38201G 1 g
D38205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D38201G Supplier TCI America Supplier No. D38201G
Only null left

Chemical Identifiers

CAS 626-41-5
Molecular Formula C6H4Br2O
Molecular Weight (g/mol) 251.905
MDL Number MFCD06411376
InChI Key PZFMWYNHJFZBPO-UHFFFAOYSA-N
PubChem CID 12280
IUPAC Name 3,5-dibromophenol
SMILES C1=C(C=C(C=C1Br)Br)O

Specifications

Melting Point 81°C
Boiling Point 122°C
Quantity 1 g
UN Number 2811
Synonym phenol, 3,5-dibromo, 3,5-dibromo-phenol, 3,5-dibromphenol, 3, 5-dibromophenol, 3-,5-dibromophenol, pubchem23831, acmc-1az7g, 3,5-dibromophenol, ksc493e8n, akos akm01584
Formula Weight 251.91
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dibromophenol