missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromophenol, 97%

Catalog No. p-7040777
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6064204 2 g
AAH6064209 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6064204 Supplier Thermo Scientific Chemicals Supplier No. H6064204
Only null left

CAS: 626-41-5 | C6H4Br2O | 251.905 g/mol

3,5-Dibromophenol is used as a pharmaceutical intermediate. It is also used to prepare aromatic polyesters and drugs for arthritis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dibromophenol is used as a pharmaceutical intermediate. It is also used to prepare aromatic polyesters and drugs for arthritis.

Solubility
Sightly soluble in water. Soluble in methanol.

Notes
Incompatible with strong oxidizing agents. Light sensitive.

Chemical Identifiers

CAS 626-41-5
Molecular Formula C6H4Br2O
Molecular Weight (g/mol) 251.905
MDL Number MFCD06411376
InChI Key PZFMWYNHJFZBPO-UHFFFAOYSA-N
PubChem CID 12280
IUPAC Name 3,5-dibromophenol
SMILES C1=C(C=C(C=C1Br)Br)O

Specifications

Melting Point 79°C to 83°C
Quantity 2 g
UN Number UN2811
Synonym phenol, 3,5-dibromo, 3,5-dibromo-phenol, 3,5-dibromphenol, 3, 5-dibromophenol, 3-,5-dibromophenol, pubchem23831, acmc-1az7g, 3,5-dibromophenol, ksc493e8n, akos akm01584
Solubility Information Sightly soluble in water. Soluble in methanol.
Formula Weight 251.9
Percent Purity 97%
Chemical Name or Material 3,5-Dibromophenol
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only