missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dibromodithieno[3,2-b:2',3'-d]thiophene 96.0+%, TCI America™
$570.40 - $570.40
Chemical Identifiers
| CAS | 502764-54-7 |
|---|---|
| Molecular Formula | C8H2Br2S3 |
| Molecular Weight (g/mol) | 354.09 |
| MDL Number | MFCD11870114 |
| InChI Key | MFLUNTNNZSCLFW-UHFFFAOYSA-N |
| PubChem CID | 11462346 |
| IUPAC Name | 5,9-dibromo-3,7,11-trithiatricyclo[6.3.0.0²,â¶]undeca-1(8),2(6),4,9-tetraene |
| SMILES | BrC1=CSC2=C1SC1=C2SC=C1Br |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D3800200MG
|
TCI America
D3800200MG |
200 mg |
Each for $570.40
|
|
|||||
Chemical Identifiers
| 502764-54-7 | |
| 354.09 | |
| MFLUNTNNZSCLFW-UHFFFAOYSA-N | |
| 5,9-dibromo-3,7,11-trithiatricyclo[6.3.0.0²,â¶]undeca-1(8),2(6),4,9-tetraene |
| C8H2Br2S3 | |
| MFCD11870114 | |
| 11462346 | |
| BrC1=CSC2=C1SC1=C2SC=C1Br |
Specifications
| 502764-54-7 | |
| Yellow | |
| MFCD11870114 | |
| MFLUNTNNZSCLFW-UHFFFAOYSA-N | |
| 5,9-dibromo-3,7,11-trithiatricyclo[6.3.0.0²,â¶]undeca-1(8),2(6),4,9-tetraene | |
| 11462346 | |
| ≥96.0% (GC) | |
| 3,5-Dibromodithieno[3,2-b:2′,3′-d]thiophene |
| 238°C | |
| C8H2Br2S3 | |
| 200 mg | |
| BrC1=CSC2=C1SC1=C2SC=C1Br | |
| 354.09 | |
| 354.09 | |
| Crystalline Powder |
Safety and Handling
TSCA : No