accessibility menu, dialog, popup

UserName

CAS RN 502764-54-7

IUPAC Name: 5,9-dibromo-3,7,11-trithiatricyclo[6.3.0.0²,⁶]undeca-1(8),2(6),4,9-tetraene
Synonyms: 5,9-dibromo-3,7,11-trithiatricyclo[6.3.0.0²,⁶]undeca-1(8),2(6),4,9-tetraene
Molecular Weight (g/mol): 354.09
Molecular Formula: C8H2Br2S3
InChi Key: MFLUNTNNZSCLFW-UHFFFAOYSA-N
SMILES: BrC1=CSC2=C1SC1=C2SC=C1Br
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results